Anti-ZBTB16 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBIZ0744-35-HRPEA 6360 DKK
USBIZ0744-35-HRP
Anti-ZBTB16 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Recommended dilution: ELISA: 1:1000 Dot Blot: 1:500
Type: Primary
Antigen: ZBTB16 (zinc finger and BTB domain containing 16)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human