Anti-VDAC2 Goat Polyclonal Antibody
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBIV2121-21EEA 4730 DKK
USBIV2121-21E
Anti-VDAC2 Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:64000; Western Blot: 1:64000; Immunochemistry: 2.5-5 ?g/ml
Type: Primary
Antigen: VDAC2 (voltage-dependent anion channel 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype:
Reactivity: Human