Anti-MSRA Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIM4692-59GEA 4730 DKK
USBIM4692-59G
Anti-MSRA Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1:2000; Immunochemistry: 1:200

Type: Primary
Antigen: MSRA (methionine sulfoxide reductase A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR