Anti-Microglobulin Mouse Monoclonal Antibody [clone: 6D585]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIM3890-03AEA 3650 DKK
USBIM3890-03A
Anti-Microglobulin Mouse Monoclonal Antibody [clone: 6D585]
Antistoffer
Recommended dilution: Flow Cytometry: 1 ?g/ml; Western Blot: 2-4?g/ml; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: Microglobulin
Clonality: monoclonal
Clone: 6D585
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR