Anti-HA tag Chicken Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBII7650-17DEA 3490 DKK
USBII7650-17D
Anti-HA tag Chicken Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:20000; Western Blot: 1:1000

Type: Tag
Antigen: HA tag (HA tag)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR