Anti-Influenza A, USSR Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBII7650-05HEA 3730 DKK
USBII7650-05H
Anti-Influenza A, USSR Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Recommended dilution: Immunofluorescence: 1:1000; Hemagglutination inhibition: 1:2500

Type: Primary
Antigen: Influenza A, USSR
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR