Anti-CYBRD1 Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9831-01BEA 3460 DKK
USBID9831-01B
Anti-CYBRD1 Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:16000; Immunochemistry: 2-5 ?g/ml

Type: Primary
Antigen: CYBRD1 (cytochrome b reductase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Pig
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR