Anti-DTX4 Rabbit Polyclonal Antibody (Biotin)
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBID9644-04-BIOTEA 6360 DKK
USBID9644-04-BIOT
Anti-DTX4 Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: DTX4 (deltex 4, E3 ubiquitin ligase)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human