Anti-DTX3L Mouse monoclonal antibody Biotin [clone: 10C19]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9644-01C-BIOTEA 0 DKK
USBID9644-01C-BIOT
Anti-DTX3L Mouse monoclonal antibody Biotin [clone: 10C19]
Type: Primary
Antigen: DTX3L (deltex 3 like, E3 ubiquitin ligase)
Clonality: monoclonal
Clone: 10C19
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR