Anti-SHFM1 Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9642-01BEA 3460 DKK
USBID9642-01B
Anti-SHFM1 Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1: 32000

Type: Primary
Antigen: SHFM1 (split hand/foot malformation (ectrodactyly) type 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR