Anti-DVL3 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9641-01DEA 3480 DKK
USBID9641-01D
Anti-DVL3 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1-3?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: DVL3 (dishevelled segment polarity protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog, Horse, Human, Monkey, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR