Anti-DVL3 Mouse Monoclonal Antibody [clone: 9G349]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9641-01CEA 3700 DKK
USBID9641-01C
Anti-DVL3 Mouse Monoclonal Antibody [clone: 9G349]
Antistoffer
Type: Primary
Antigen: DVL3 (dishevelled segment polarity protein 3)
Clonality: monoclonal
Clone: 9G349
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR