Anti-DSCAM Mouse Monoclonal Antibody [clone: 7H209]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9635-50EA 4320 DKK
USBID9635-50
Anti-DSCAM Mouse Monoclonal Antibody [clone: 7H209]
Antistoffer
Type: Primary
Antigen: DSCAM (Down syndrome cell adhesion molecule)
Clonality: monoclonal
Clone: 7H209
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR