Anti-DRAXIN Sheep Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9392-02EA 4590 DKK
USBID9392-02
Anti-DRAXIN Sheep Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1 ?g/ml; ELISA: 0.5-1 ?g/ml

Type: Primary
Antigen: DRAXIN (dorsal inhibitory axon guidance protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR