Anti-DRAM1 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9370-02CEA 5140 DKK
USBID9370-02C
Anti-DRAM1 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 0.5-1 ?g/ml; Immunochemistry: 2.5-?g/ml

Type: Primary
Antigen: DRAM1 (DNA-damage regulated autophagy modulator 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR