Anti-STK17B Rabbit Polyclonal Antibody
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBID9335-03CEA 5040 DKK
USBID9335-03C
Anti-STK17B Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Immunochemistry: 1:50
Type: Primary
Antigen: STK17B (serine/threonine kinase 17b)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human