Anti-STK17A Rabbit Polyclonal Antibody (Biotin)

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9334-01G-BIOTEA 6360 DKK
USBID9334-01G-BIOT
Anti-STK17A Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: STK17A (serine/threonine kinase 17a)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR