Anti-STK17A Mouse Monoclonal Antibody [clone: 8C120]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9334-01BEA 2530 DKK
USBID9334-01B
Anti-STK17A Mouse Monoclonal Antibody [clone: 8C120]
Antistoffer
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: STK17A (serine/threonine kinase 17a)
Clonality: monoclonal
Clone: 8C120
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR