Anti-TNFRSF21 Rat Monoclonal Antibody [clone: 6D361]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9300-62CEA 4100 DKK
USBID9300-62C
Anti-TNFRSF21 Rat Monoclonal Antibody [clone: 6D361]
Antistoffer
Type: Primary
Antigen: TNFRSF21 (tumor necrosis factor receptor superfamily, member 21)
Clonality: monoclonal
Clone: 6D361
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR