Anti-TNFRSF21 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9300-61CEA 5190 DKK
USBID9300-61C
Anti-TNFRSF21 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1 ?g/ml; Immunochemistry: 3?g/ml

Type: Primary
Antigen: TNFRSF21 (tumor necrosis factor receptor superfamily, member 21)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR