Anti-TNFRSF10B Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9300-50GEA 3310 DKK
USBID9300-50G
Anti-TNFRSF10B Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 5 ?g/ml

Type: Primary
Antigen: TNFRSF10B (tumor necrosis factor receptor superfamily, member 10b)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR