Anti-TNFRSF10A Mouse Monoclonal Antibody [clone: 10B1165]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9300-30ZEA 4290 DKK
USBID9300-30Z
Anti-TNFRSF10A Mouse Monoclonal Antibody [clone: 10B1165]
Antistoffer
Recommended dilution: Flow Cytometry: 1:50-1:100;

Type: Primary
Antigen: TNFRSF10A (tumor necrosis factor receptor superfamily, member 10a)
Clonality: monoclonal
Clone: 10B1165
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR