Anti-DPH2 Mouse monoclonal antibody PE (Phycoerythrin) [clone: 10B2758]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9150-75-PEEA 0 DKK
USBID9150-75-PE
Anti-DPH2 Mouse monoclonal antibody PE (Phycoerythrin) [clone: 10B2758]
Type: Primary
Antigen: DPH2 (DPH2 homolog (S. cerevisiae))
Clonality: monoclonal
Clone: 10B2758
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR