Anti-DPH2 Mouse monoclonal antibody HRP (Horseradish Peroxidase) [clone: 10B2758]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9150-75-HRPEA 0 DKK
USBID9150-75-HRP
Anti-DPH2 Mouse monoclonal antibody HRP (Horseradish Peroxidase) [clone: 10B2758]
Type: Primary
Antigen: DPH2 (DPH2 homolog (S. cerevisiae))
Clonality: monoclonal
Clone: 10B2758
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR