Anti-DPF2 Mouse monoclonal antibody HRP (Horseradish Peroxidase) [clone: 10B2756]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9150-70B-HRPEA 0 DKK
USBID9150-70B-HRP
Anti-DPF2 Mouse monoclonal antibody HRP (Horseradish Peroxidase) [clone: 10B2756]
Type: Primary
Antigen: DPF2 (D4, zinc and double PHD fingers family 2)
Clonality: monoclonal
Clone: 10B2756
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR