Anti-DCX Rabbit Polyclonal Antibody (FITC (Fluorescein))

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9080-01A-FITCEA 4620 DKK
USBID9080-01A-FITC
Anti-DCX Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-1:100; Immunochemistry: 1:10-1:50
Do not freeze!

Type: Primary
Antigen: DCX (doublecortin)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR