Anti-DOT1L Mouse Monoclonal Antibody [clone: 11n62]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBID9056-01F1EA 5540 DKK
USBID9056-01F1
Anti-DOT1L Mouse Monoclonal Antibody [clone: 11n62]
Antistoffer
Recommended dilution: Immunochemistry: 8-25 ?g/ml; Immunoprecipitation: 25 ?g/ml
Type: Primary
Antigen: DOT1L (DOT1-like histone H3K79 methyltransferase)
Clonality: monoclonal
Clone: 11n62
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human