Anti-DOT1L Rabbit Polyclonal Antibody (Biotin)

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9056-01E-BIOTEA 4620 DKK
USBID9056-01E-BIOT
Anti-DOT1L Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: DOT1L (DOT1-like histone H3K79 methyltransferase)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR