Anti-Dopamine Transporter Rabbit Polyclonal Antibody
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBID9020-30AEA 5540 DKK
USBID9020-30A
Anti-Dopamine Transporter Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Immunochemistry: 1:1000; Western Blot: 1:1000
Type: Primary
Antigen: Dopamine Transporter
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Monkey