Anti-Dopamine Transporter Rat Monoclonal Antibody [clone: 10B2323]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9020-01JEA 6450 DKK
USBID9020-01J
Anti-Dopamine Transporter Rat Monoclonal Antibody [clone: 10B2323]
Antistoffer
Recommended dilution: Immunochemistry: 1:1000-1:10000

Type: Primary
Antigen: Dopamine Transporter
Clonality: monoclonal
Clone: 10B2323
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR