Anti-Dopamine Receptor, D1b Mouse Monoclonal Antibody [clone: 8J1312]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9015-01EA 3310 DKK
USBID9015-01
Anti-Dopamine Receptor, D1b Mouse Monoclonal Antibody [clone: 8J1312]
Antistoffer
Type: Primary
Antigen: Dopamine Receptor, D1b
Clonality: monoclonal
Clone: 8J1312
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR