Anti-Dopa Decarboxylase Goat Polyclonal Antibody (Biotin)

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9013-05EA 5140 DKK
USBID9013-05
Anti-Dopa Decarboxylase Goat Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: Dopa Decarboxylase
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR