Anti-Dopamine Decarboxylase Mouse Monoclonal Antibody [clone: 7H245]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9013-01FEA 5680 DKK
USBID9013-01F
Anti-Dopamine Decarboxylase Mouse Monoclonal Antibody [clone: 7H245]
Antistoffer
Recommended dilution: Western Blot: 2?g/ml

Type: Primary
Antigen: Dopamine Decarboxylase
Clonality: monoclonal
Clone: 7H245
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR