Anti-Dopamine Beta Hydroxylase Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9010-13EA 3700 DKK
USBID9010-13
Anti-Dopamine Beta Hydroxylase Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Dopamine Beta Hydroxylase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR