Anti-Dopamine Beta Hydroxylase Sheep Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9010-03EA 3410 DKK
USBID9010-03
Anti-Dopamine Beta Hydroxylase Sheep Polyclonal Antibody
Antistoffer
Recommended dilution: Immunochemistry: 1:5000-50000; Western Blot: 1:5000-1:10000

Type: Primary
Antigen: Dopamine Beta Hydroxylase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR