Anti-Dopamine Beta Hydroxylase Sheep Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9010-02AEA 5530 DKK
USBID9010-02A
Anti-Dopamine Beta Hydroxylase Sheep Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: Dopamine Beta Hydroxylase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Sheep
Isotype: IgG
Reactivity: Human, Monkey, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR