Anti-Dopamine Receptor, D5 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9000-23FEA 3040 DKK
USBID9000-23F
Anti-Dopamine Receptor, D5 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Dopamine Receptor, D5
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR