Anti-Dopamine Receptor, D3 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9000-15BEA 3060 DKK
USBID9000-15B
Anti-Dopamine Receptor, D3 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1:500; Immunochemistry: 1:5000

Type: Primary
Antigen: Dopamine Receptor, D3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR