Anti-Dopamine Receptor, D2s Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9000-14AEA 3060 DKK
USBID9000-14A
Anti-Dopamine Receptor, D2s Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1:800; Immunochemistry: 1:500

Type: Primary
Antigen: Dopamine Receptor, D2s
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR