Anti-Dopamine Receptor, D2L Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9000-09BEA 4730 DKK
USBID9000-09B
Anti-Dopamine Receptor, D2L Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Immunochemistry: 20 ?g/ml

Type: Primary
Antigen: Dopamine Receptor, D2L
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR