Anti-DRD2 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9000-06LEA 3040 DKK
USBID9000-06L
Anti-DRD2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: DRD2 (dopamine receptor D2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR