Anti-DRD2 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9000-06GEA 2800 DKK
USBID9000-06G
Anti-DRD2 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Immunochemistry: 1:5000; Western Blot: 1:800

Type: Primary
Antigen: DRD2 (dopamine receptor D2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR