Anti-DRD1 Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID9000-05JEA 2750 DKK
USBID9000-05J
Anti-DRD1 Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:64000

Type: Primary
Antigen: DRD1 (dopamine receptor D1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Dog, Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR