Anti-DOLK Rabbit Polyclonal Antibody (Biotin)

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8085-85-BIOTEA 4620 DKK
USBID8085-85-BIOT
Anti-DOLK Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: DOLK (dolichol kinase)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR