Anti-DOK5 Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8080-12BEA 4120 DKK
USBID8080-12B
Anti-DOK5 Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:16000; Western Blot: 1-3?g/ml

Type: Primary
Antigen: DOK5 (docking protein 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR