Anti-DOCK2 Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8080-05MEA 4590 DKK
USBID8080-05M
Anti-DOCK2 Goat Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot:1g/ml

Type: Primary
Antigen: DOCK2 (dedicator of cytokinesis 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Goat, Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR