Anti-DOK2 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8080-02EEA 2840 DKK
USBID8080-02E
Anti-DOK2 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot:1:500-1:1000; Immunochemistry:1:50-1:100

Type: Primary
Antigen: DOK2 (docking protein 2, 56kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR