Anti-Dok, p62 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8080-01AEA 4600 DKK
USBID8080-01A
Anti-Dok, p62 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:10-1:50

Type: Primary
Antigen: Dok, p62
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR