Anti-ANO1 Rabbit Monoclonal Antibody [clone: 10A20]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8079-35EA 3340 DKK
USBID8079-35
Anti-ANO1 Rabbit Monoclonal Antibody [clone: 10A20]
Antistoffer
Recommended dilution: Immunochemistry: 1:50

Type: Primary
Antigen: ANO1 (anoctamin 1, calcium activated chloride channel)
Clonality: monoclonal
Clone: 10A20
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR