Anti-DOCK4 Mouse monoclonal antibody Biotin [clone: 10B2754]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8077-30-BIOTEA 0 DKK
USBID8077-30-BIOT
Anti-DOCK4 Mouse monoclonal antibody Biotin [clone: 10B2754]
Type: Primary
Antigen: DOCK4 (dedicator of cytokinesis 4)
Clonality: monoclonal
Clone: 10B2754
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR